Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CNGC6
Entrez Gene ID
816931
Gene Name
cyclic nucleotide-gated channel 6
Gene Aliases
AT2G23980, ATCNGC6, CYCLIC NUCLEOTIDE GATED CHANNEL 6, T29E15.18, T29E15_18, cyclic nucleotide-gated channel 6
Chromosome Location
Chr.2: 10201104 - 10205422 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001335878.1 NP_001318278.1 4 - 5 877 97
NM_001335879.1 NP_001323882.1 3 - 4 1224 97
NM_001335880.1 NP_001323881.1 2 - 3 1179 97
NM_001335881.1 NP_001323885.1 3 - 4 1224 97
NM_001335882.1 NP_001323879.1 2 - 3 1179 97
NM_001335883.1 NP_001323880.1 1 - 2 1024 97
NM_001335884.1 NP_001323886.1 4 - 5 877 97
NM_001335885.1 NP_001323884.1 3 - 4 832 97
NM_001335886.1 NP_001323883.1 3 - 4 832 97
NM_127960.3 NP_565560.1 3 - 4 756 97
GenBank mRNA Y17914.1 - - 656 97

View Full Product Details