Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CNGC5
Entrez Gene ID
835905
Gene Name
cyclic nucleotide gated channel 5
Gene Aliases
AT5G57940, ATCNGC5, MTI20.20, MTI20_20, cyclic nucleotide gated channel 5
Chromosome Location
Chr.5: 23456565 - 23460621 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_125179.5 NP_200602.2 4 - 5 1020 88
NM_180878.2 NP_851209.1 4 - 5 1149 88
NM_203224.1 NP_974953.1 3 - 4 797 88
GenBank mRNA AK317271.1 - 4 - 5 1012 88
AY091133.1 - 4 - 5 1149 88
AY114053.1 - 3 - 4 818 88
Y17913.1 - - 797 88

View Full Product Details