Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MKP1
- Entrez Gene ID
-
824693
- Gene Name
- mitogen-activated protein kinase phosphatase 1
- Gene Aliases
-
ARABIDOPSIS MITOGEN-ACTIVATED PROTEIN KINASE PHOSPHATASE 1, AT3G55270, ATMKP1, mitogen-activated protein kinase phosphatase 1
- Chromosome Location
-
Chr.3: 20496081 - 20500019 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_115385.7 |
NP_567018.5 |
3 - 4 |
2829 |
77 |
View Full Product Details