Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BGL2
Entrez Gene ID
824893
Gene Name
beta-1,3-glucanase 2
Gene Aliases
''beta-1, 3-GLUCANASE, 3-GLUCANASE 2, 3-glucanase 2, 3-glucanase 2'', AT3G57260, AtBG2, AtPR2, BETA-1, BG2, PATHOGENESIS-RELATED PROTEIN 2, PR-2, PR2, beta-1
Chromosome Location
Chr.3: 21188397 - 21189967 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001339849.1 NP_001325845.1 1 - 2 233 71
NM_115586.2 NP_191285.1 1 - 2 125 71
GenBank mRNA AY099668.1 - 1 - 2 125 71
AY128847.1 - 1 - 2 88 71

View Full Product Details