Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DEG8
Entrez Gene ID
833979
Gene Name
Trypsin family protein with PDZ domain-containing protein
Gene Aliases
AT5G39830, DEG PROTEASE 8, DEGP8, K13H13.1, K13H13_1, degradation of periplasmic proteins 8
Chromosome Location
Chr.5: 15942764 - 15945897 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_123346.5 NP_568575.1 7 - 8 776 81
NM_203134.2 NP_974863.1 7 - 8 743 81
GenBank mRNA AY056381.1 - 7 - 8 720 81
AY090319.1 - 7 - 8 657 81

View Full Product Details