Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SHM7
Entrez Gene ID
840543
Gene Name
serine hydroxymethyltransferase 7
Gene Aliases
AT1G36370, F7F23.9, F7F23_9, serine hydroxymethyltransferase 7
Chromosome Location
Chr.1: 13696042 - 13698739 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_103323.3 NP_564473.1 2 - 3 1620 64
GenBank mRNA BT002738.1 - 2 - 3 1457 64

View Full Product Details