Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TLP6
Entrez Gene ID
841130
Gene Name
tubby like protein 6
Gene Aliases
AT1G47270, AtTLP6, F8G22.1, F8G22_1, tubby like protein 6
Chromosome Location
Chr.1: 17326530 - 17328772 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198230.2 NP_001185159.1 5 - 6 915 112
NM_001333268.1 NP_001322089.1 4 - 5 674 112
NM_103621.2 NP_175160.2 5 - 6 892 112
GenBank mRNA AF487268.1 - 3 - 4 545 112
AK228930.1 - 5 - 6 892 112

View Full Product Details