Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SHP1
Entrez Gene ID
825047
Gene Name
K-box region and MADS-box transcription factor family protein
Gene Aliases
AGAMOUS-like 1, AGL1, AT3G58780, SHATTERPROOF 1
Chromosome Location
Chr.3: 21738160 - 21742517 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084842.1 NP_001078311.1 3 - 4 613 82
NM_001203201.2 NP_001190130.1 5 - 6 963 82
NM_001339937.1 NP_001325709.1 3 - 4 934 82
NM_001339938.1 NP_001325710.1 2 - 3 580 82
NM_115740.3 NP_191437.1 3 - 4 934 82
GenBank mRNA AY086196.1 - 3 - 4 634 82
DQ446777.1 - 2 - 3 292 82
JQ973091.1 - 2 - 3 313 82
M55550.1 - 2 - 3 319 82

View Full Product Details