Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SHP1
Entrez Gene ID
825047
Gene Name
K-box region and MADS-box transcription factor family protein
Gene Aliases
AGAMOUS-like 1, AGL1, AT3G58780, SHATTERPROOF 1
Chromosome Location
Chr.3: 21738160 - 21742517 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084842.1 NP_001078311.1 6 - 7 813 75
NM_001203201.2 NP_001190130.1 8 - 9 1163 75
NM_001339937.1 NP_001325709.1 5 - 6 1233 75
NM_001339938.1 NP_001325710.1 5 - 6 780 75
NM_115740.3 NP_191437.1 6 - 7 1134 75
GenBank mRNA AY086196.1 - 6 - 7 834 75
DQ446777.1 - 5 - 6 492 75
JQ973091.1 - 5 - 6 513 75
M55550.1 - 5 - 6 519 75

View Full Product Details