Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GAUT15
Entrez Gene ID
825048
Gene Name
galacturonosyltransferase 15
Gene Aliases
AT3G58790, galacturonosyltransferase 15
Chromosome Location
Chr.3: 21742071 - 21746324 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_115741.3 NP_191438.2 2 - 3 633 75
GenBank mRNA AY099698.1 - 2 - 3 633 75
AY128894.1 - 2 - 3 281 75
KJ138783.1 - 2 - 3 281 75

View Full Product Details