Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GCN2
Entrez Gene ID
825110
Gene Name
protein kinase family protein
Gene Aliases
ARABIDOPSIS THALIANA GENERAL CONTROL NON-REPRESSIBLE 2, AT3G59410, ATGCN2, GENERAL CONTROL NON-REPRESSIBLE 2
Chromosome Location
Chr.3: 21948851 - 21959410 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203206.1 NP_001190135.1 11 - 12 1873 76
NM_001339969.1 NP_001326041.1 11 - 12 2740 76
NM_001339970.1 NP_001326042.1 10 - 11 1882 76
NM_001339971.1 NP_001326040.1 10 - 11 1653 76
NM_115803.3 NP_191500.2 10 - 11 1772 76
GenBank mRNA AJ459823.1 - 10 - 11 1420 76

View Full Product Details