Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GCN2
Entrez Gene ID
825110
Gene Name
protein kinase family protein
Gene Aliases
ARABIDOPSIS THALIANA GENERAL CONTROL NON-REPRESSIBLE 2, AT3G59410, ATGCN2, GENERAL CONTROL NON-REPRESSIBLE 2
Chromosome Location
Chr.3: 21948851 - 21959410 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203206.1 NP_001190135.1 22 - 23 3406 67
NM_001339969.1 NP_001326041.1 22 - 23 4273 67
NM_001339970.1 NP_001326042.1 21 - 22 3415 67
NM_001339971.1 NP_001326040.1 21 - 22 3186 67
NM_115803.3 NP_191500.2 21 - 22 3305 67
GenBank mRNA AJ459823.1 - 21 - 22 2953 67

View Full Product Details