Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
U2B''
Entrez Gene ID
817576
Gene Name
U2 small nuclear ribonucleoprotein B
Gene Aliases
AT2G30260, T9D9.7, T9D9_7, U2 small nuclear ribonucleoprotein B
Chromosome Location
Chr.2: 12905345 - 12907665 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_128579.4 NP_180585.1 4 - 5 660 80
GenBank mRNA AK227609.1 - 4 - 5 504 80
BT003068.1 - 4 - 5 429 80

View Full Product Details