Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FTSH6
Entrez Gene ID
831377
Gene Name
FTSH protease 6
Gene Aliases
AT5G15250, ATFTSH6, F8M21.140, F8M21_140, FTSH protease 6
Chromosome Location
Chr.5: 4950359 - 4952803 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_121529.2 NP_568311.2 3 - 4 1477 76
GenBank mRNA DQ446953.1 - 3 - 4 1477 76

View Full Product Details