Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ASP2
- Entrez Gene ID
-
832075
- Gene Name
- aspartate aminotransferase 2
- Gene Aliases
-
AAT2, ASPARTATE AMINOTRANSFERASE, ASPARTATE AMINOTRANSFERASE 2, ASPAT, AT5G19550, CYTOPLASMIC ISOZYME 1, T20D1.70, T20D1_70, aspartate aminotransferase 2
- Chromosome Location
-
Chr.5: 6597971 - 6601971 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_121960.5 |
NP_197456.1 |
3 - 4 |
417 |
100 |
View Full Product Details