Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ZIP3
- Entrez Gene ID
-
817787
- Gene Name
- zinc transporter 3 precursor
- Gene Aliases
-
AT2G32270, T32F6.21, T32F6_21, zinc transporter 3 precursor
- Chromosome Location
-
Chr.2: 13704068 - 13706928 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_128786.4 |
NP_180786.1 |
1 - 2 |
768 |
113 |
View Full Product Details