Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PAP13
Entrez Gene ID
817838
Gene Name
purple acid phosphatase 13
Gene Aliases
AT2G32770, ATPAP13, F24L7.9, F24L7_9, PURPLE ACID PHOSPHATASE 13, purple acid phosphatase 13
Chromosome Location
Chr.2: 13895825 - 13898389 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_128837.1 NP_180836.1 2 - 3 833 118
NM_179867.1 NP_850198.1 1 - 2 1125 118
NM_201856.4 NP_973585.1 2 - 3 846 118
GenBank mRNA AF492665.1 - 2 - 3 833 118

View Full Product Details