The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198270.1 | NP_001185199.1 | 12 - 13 | 1490 | 101 |
NM_001333494.1 | NP_001322512.1 | 10 - 11 | 3389 | 101 | |
NM_001333495.1 | NP_001322511.1 | 12 - 13 | 1511 | 101 | |
NM_104047.4 | NP_564595.1 | 11 - 12 | 3118 | 101 | |
NM_202274.2 | NP_974003.1 | 10 - 11 | 1412 | 101 | |
GenBank mRNA | AK230310.1 | - | 11 - 12 | 2898 | 101 |
AY120756.1 | - | 10 - 11 | 1413 | 101 | |
BT000109.1 | - | 10 - 11 | 1171 | 101 |