Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RLK4
Entrez Gene ID
828079
Gene Name
receptor-like protein kinase 4
Gene Aliases
AT4G00340, A_IG005I10.19, A_IG005I10_19, receptor-like protein kinase 4
Chromosome Location
Chr.4: 148612 - 151739 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001340241.1 NP_001329840.1 2 - 3 2031 84
NM_116257.5 NP_567172.4 1 - 2 2440 84

View Full Product Details