Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PIPK10
Entrez Gene ID
828066
Gene Name
phosphatidylinositol phosphate kinase 10
Gene Aliases
AT4G01190, ATPIPK10, F2N1.9, F2N1_9, phosphatidylinositol phosphate kinase 10
Chromosome Location
Chr.4: 504135 - 506403 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001340304.1 NP_001329151.1 3 - 4 661 142
NM_001340305.1 NP_001329152.1 4 - 5 545 142
NM_116349.1 NP_192028.1 3 - 4 389 142

View Full Product Details