Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GLY3
Entrez Gene ID
841793
Gene Name
glyoxalase II 3
Gene Aliases
AT1G53580, ETHE1, ETHE1-LIKE, ETHYLMALONIC ENCEPHALOPATHY PROTEIN1, F22G10.9, GLX2-3, GLYOXALASE 2-3, T3F20.11, T3F20_11, glyoxalase II 3, glyoxalase II 3
Chromosome Location
Chr.1: 19991271 - 19993458 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_104236.5 NP_564636.2 5 - 6 860 83
NM_202289.3 NP_974018.3 5 - 6 736 83

View Full Product Details