Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
XIE
Entrez Gene ID
841898
Gene Name
Myosin family protein with Dil domain-containing protein
Gene Aliases
AT1G54560, ATXIE, MYOSIN XI E, T22H22.1, T22H22_1
Chromosome Location
Chr.1: 20371469 - 20380156 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001333655.1 NP_001320715.1 29 - 30 3957 103
NM_104334.2 NP_175858.1 28 - 29 3681 103

View Full Product Details