Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NOP56
Entrez Gene ID
842063
Gene Name
NOP56-like pre RNA processing ribonucleoprotein
Gene Aliases
AT1G56110, NOP56-LIKE PROTEIN, T6H22.10, T6H22_10, homolog of nucleolar protein NOP56
Chromosome Location
Chr.1: 20984273 - 20986952 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_104489.4 NP_176007.1 1 - 2 262 105
GenBank mRNA AF302492.1 - 1 - 2 203 105
AY039541.1 - 1 - 2 257 105
AY087080.1 - 1 - 2 259 105
AY102151.1 - 1 - 2 203 105

View Full Product Details