Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BCA6
Entrez Gene ID
842185
Gene Name
beta carbonic anhydrase 6
Gene Aliases
A. THALIANA BETA CARBONIC ANHYDRASE 6, AT1G58180, ATBCA6, T15M6.18, beta carbonic anhydrase 6
Chromosome Location
Chr.1: 21537577 - 21540038 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036129.3 NP_001031206.1 8 - 9 977 162
NM_001198330.1 NP_001185259.1 9 - 10 883 162
NM_001333817.1 NP_001320705.1 8 - 9 963 162
NM_001333818.1 NP_001320704.1 7 - 8 673 162
NM_104599.5 NP_176114.2 9 - 10 1007 162
GenBank mRNA AK316811.1 - 8 - 9 859 162

View Full Product Details