Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RXF12
Entrez Gene ID
842206
Gene Name
glycosyl hydrolase family 10 protein / carbohydrate-binding domain-containing protein
Gene Aliases
ARABIDOPSIS THALIANA XYLANASE 1, AT1G58370, ATXYN1, F9K23.10
Chromosome Location
Chr.1: 21684263 - 21688359 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001333829.1 NP_001323263.1 6 - 7 1443 104
NM_104617.4 NP_176133.1 6 - 7 1140 104
GenBank mRNA AB008015.1 - 6 - 7 1122 104

View Full Product Details