Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CW9
Entrez Gene ID
842252
Gene Name
Disease resistance protein (CC-NBS-LRR class) family
Gene Aliases
AT1G59620, T30E16.18, T30E16_18
Chromosome Location
Chr.1: 21902264 - 21905717 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001333858.1 NP_001319272.1 2 - 3 1013 101
NM_001333859.1 NP_001320323.1 2 - 3 993 101

View Full Product Details