Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNL5
Entrez Gene ID
842283
Gene Name
SIN3-like 5
Gene Aliases
AT1G59890, F23H11.20, F23H11_20, SIN3-like 5
Chromosome Location
Chr.1: 22043643 - 22051114 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198338.1 NP_001185267.1 18 - 19 3665 96
NM_001198339.2 NP_001185268.1 18 - 19 3878 96
NM_001198340.2 NP_001185269.1 18 - 19 3846 96
NM_104681.6 NP_176197.3 18 - 19 3863 96

View Full Product Details