Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PANK1
- Entrez Gene ID
-
842339
- Gene Name
- pantothenate kinase 1
- Gene Aliases
-
AT1G60440, ATCOAA, ATPANK1, T13D8.31, T13D8_31, pantothenate kinase 1
- Chromosome Location
-
Chr.1: 22266416 - 22269258 on Build TAIR10.1
- Gene Symbol
- FLA2
- Entrez Gene ID
-
826885
- Gene Name
- FASCICLIN-like arabinogalactan 2
- Gene Aliases
-
AT4G12730, FASCICLIN-like arabinogalactan 2, T20K18.80, T20K18_80
- Chromosome Location
-
Chr.4: 7491227 - 7493080 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK230362.1 |
- |
- |
938 |
138 |
View Full Product Details