Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ABC4
Entrez Gene ID
842354
Gene Name
UbiA prenyltransferase family protein
Gene Aliases
1, 4-DIHYDROXY-2-NAPTHOIC ACID PHYTYLTRANSFERASE, ABERRANT CHLOROPLAST DEVELOPMENT 4, AT1G60600, F8A5.36, F8A5_36
Chromosome Location
Chr.1: 22324437 - 22327359 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001124046.2 NP_001117518.1 8 - 9 1268 113
NM_104743.3 NP_176259.2 8 - 9 1260 113
GenBank mRNA AK227267.1 - 8 - 9 1191 113

View Full Product Details