Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KAS III
Entrez Gene ID
842561
Gene Name
3-ketoacyl-acyl carrier protein synthase III
Gene Aliases
3-KETOACYL-ACYL CARRIER PROTEIN SYNTHASE III, 3-ketoacyl-acyl carrier protein synthase III, AT1G62640, T3P18.20, T3P18_20
Chromosome Location
Chr.1: 23192146 - 23195103 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036144.2 NP_001031221.1 2 - 3 460 85
NM_104942.4 NP_176452.1 1 - 2 545 85
GenBank mRNA AY063804.1 - 1 - 2 405 85
AY091275.1 - 1 - 2 189 85

View Full Product Details