The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001084295.2 | NP_001077764.1 | 1 - 1 | 752 | 137 |
NM_104981.4 | NP_176491.1 | 2 - 2 | 618 | 137 |