Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SKU5
Entrez Gene ID
826851
Gene Name
Cupredoxin superfamily protein
Gene Aliases
AT4G12420, T1P17.10, T1P17_10
Chromosome Location
Chr.4: 7349187 - 7353135 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203775.2 NP_001190704.1 3 - 4 784 118
NM_117312.4 NP_192979.1 3 - 4 766 118
GenBank mRNA AF439406.1 - 3 - 4 517 118

View Full Product Details