Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SKU5
Entrez Gene ID
826851
Gene Name
Cupredoxin superfamily protein
Gene Aliases
AT4G12420, T1P17.10, T1P17_10
Chromosome Location
Chr.4: 7349187 - 7353135 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203775.2 NP_001190704.1 5 - 6 1286 88
NM_117312.4 NP_192979.1 5 - 6 1268 88
GenBank mRNA AF439406.1 - 5 - 6 1019 88

View Full Product Details