Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPA
Entrez Gene ID
818088
Gene Name
root and pollen arfgap
Gene Aliases
AGD10, AT2G35210, MATERNAL EFFECT EMBRYO ARREST 28, MEE28, T4C15.12, T4C15_12, root and pollen arfgap
Chromosome Location
Chr.2: 14836062 - 14838151 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_129074.3 NP_565801.1 2 - 3 331 67
NM_201874.2 NP_973603.1 2 - 3 331 67
GenBank mRNA AK229368.1 - 2 - 3 279 67
AY085599.1 - 2 - 3 331 67

View Full Product Details