Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RP2
Entrez Gene ID
821293
Gene Name
PPDK regulatory protein 2
Gene Aliases
AT3G01200, AtRP2, PPDK regulatory protein 2, T4P13.11, T4P13_11
Chromosome Location
Chr.3: 69468 - 71362 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_110986.4 NP_186769.1 1 - 2 399 83

View Full Product Details