Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AAE15
Entrez Gene ID
827043
Gene Name
acyl-activating enzyme 15
Gene Aliases
AT4G14070, DL3075C, FCAALL.81, acyl-activating enzyme 15
Chromosome Location
Chr.4: 8111944 - 8118188 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_117482.4 NP_193143.2 1 - 2 351 76
GenBank mRNA AY062804.1 - 1 - 2 320 76
BT010332.1 - 1 - 2 202 76
EF093797.1 - 1 - 2 202 76

View Full Product Details