Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATSUV3
Entrez Gene ID
827134
Gene Name
ATP-dependent RNA helicase
Gene Aliases
AT4G14790, DL3435C, EDA15, FCAALL.309, SUV3, embryo sac development arrest 15, suppressor of Var 3
Chromosome Location
Chr.4: 8495701 - 8500237 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001340972.1 NP_001328228.1 3 - 4 666 75
NM_117564.4 NP_193215.2 3 - 4 751 75
GenBank mRNA AJ132843.1 - 3 - 4 364 75
AY139995.1 - 3 - 4 666 75
BT008910.1 - 3 - 4 343 75

View Full Product Details