Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATG8F
Entrez Gene ID
827351
Gene Name
Ubiquitin-like superfamily protein
Gene Aliases
AT4G16520, DL4285C, FCAALL.383, autophagy 8f
Chromosome Location
Chr.4: 9306339 - 9308538 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_117751.4 NP_567504.1 3 - 4 294 112
NM_179064.2 NP_849395.1 2 - 3 419 112
GenBank mRNA AB073180.1 - 3 - 4 171 112
AY070464.1 - 3 - 4 186 112
AY091667.1 - 2 - 3 90 112

View Full Product Details