Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OFP5
Entrez Gene ID
827617
Gene Name
ovate family protein 5
Gene Aliases
AT4G18830, ATOFP5, F13C5.5, F13C5_5, ovate family protein 5
Chromosome Location
Chr.4: 10336566 - 10338674 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_117999.5 NP_193618.2 1 - 1 1755 117
GenBank mRNA AY063996.1 - 1 - 1 918 117
AY096608.1 - 1 - 1 872 117

View Full Product Details