Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
URE
Entrez Gene ID
843076
Gene Name
urease
Gene Aliases
AT1G67550, F12B7.10, F12B7_10, urease
Chromosome Location
Chr.1: 25312660 - 25317495 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105422.4 NP_176922.1 4 - 5 532 147

View Full Product Details