Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NUDX1
Entrez Gene ID
843207
Gene Name
nudix hydrolase 1
Gene Aliases
ARABIDOPSIS THALIANA NUDIX HYDROLASE HOMOLOG 1, AT1G68760, ATNUDT1, ATNUDX1, F14K14.13, F14K14_13, NUDIX HYDROLASE 1, nudix hydrolase 1, nudix hydrolase 1
Chromosome Location
Chr.1: 25828890 - 25829813 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105549.4 NP_177044.1 1 - 2 335 74
GenBank mRNA AK117564.1 - 1 - 2 151 74
AK176885.1 - 1 - 2 193 74
AY088162.1 - 1 - 2 148 74
BT005039.1 - 1 - 2 135 74

View Full Product Details