Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FIP1
Entrez Gene ID
815154
Gene Name
FRIGIDA interacting protein 1
Gene Aliases
AT2G06005, FRIGIDA interacting protein 1
Chromosome Location
Chr.2: 2332317 - 2336257 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001335289.1 NP_001318204.1 13 - 14 1224 70
NM_001335290.1 NP_001324184.1 12 - 13 1044 70
NM_001335291.1 NP_001324181.1 12 - 13 1181 70
NM_001335292.1 NP_001324182.1 12 - 13 1177 70
NM_001335293.1 NP_001324183.1 13 - 14 1096 70
NM_126614.3 NP_565332.1 13 - 14 1098 70
GenBank mRNA AY086482.1 - 13 - 14 998 70

View Full Product Details