Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SRS
Entrez Gene ID
837734
Gene Name
Seryl-tRNA synthetase
Gene Aliases
AT1G11870, ATSRS, F12F1.29, F12F1_29, OVA7, SERYL-TRNA SYNTHETASE, Seryl-tRNA synthetase, ovule abortion 7
Chromosome Location
Chr.1: 4003757 - 4007015 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001332005.1 NP_001321932.1 2 - 3 456 101
NM_001332006.1 NP_001321931.1 2 - 3 456 101
NM_101058.2 NP_172651.1 2 - 3 401 101
NM_179316.3 NP_849647.1 2 - 3 496 101
NM_202083.1 NP_973812.1 2 - 3 407 101
GenBank mRNA AY091115.1 - 2 - 3 408 101
AY122978.1 - 2 - 3 358 101

View Full Product Details