Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PEX12
Entrez Gene ID
819602
Gene Name
peroxin-12
Gene Aliases
ABERRANT PEROXISOME MORPHOLOGY 4, APM4, AT3G04460, ATPEX12, PEROXIN-12, T27C4.11, T27C4_11, peroxin-12
Chromosome Location
Chr.3: 1186440 - 1189508 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001337528.1 NP_001325637.1 3 - 4 515 108
NM_111317.4 NP_187096.2 3 - 4 559 108

View Full Product Details