Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- NIP7;1
- Entrez Gene ID
-
819783
- Gene Name
- NOD26-like intrinsic protein 7;1
- Gene Aliases
-
AT3G06100, F28L1.3, F28L1_3, NLM6, NLM8, NOD26-LIKE MIP 6, NOD26-LIKE MIP 8, NOD26-like intrinsic protein 7;1
- Chromosome Location
-
Chr.3: 1841168 - 1843196 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_111485.3 |
NP_566271.1 |
1 - 2 |
408 |
100 |
GenBank mRNA |
DQ446638.1 |
- |
1 - 2 |
146 |
100 |
View Full Product Details