Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- LCR22
- Entrez Gene ID
-
829049
- Gene Name
- low-molecular-weight cysteine-rich 22
- Gene Aliases
-
AT4G29280, F17A13.100, F17A13_100, low-molecular-weight cysteine-rich 22
- Chromosome Location
-
Chr.4: 14427882 - 14428265 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_119072.2 |
NP_194657.1 |
1 - 2 |
43 |
80 |
View Full Product Details