Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VTC5
Entrez Gene ID
835603
Gene Name
GDP-L-galactose phosphorylase VITAMIN C DEFECTIVE 5 (VTC5)
Gene Aliases
AT5G55120, MCO15.7, MCO15_7, VITAMIN C DEFECTIVE 5
Chromosome Location
Chr.5: 22369007 - 22371867 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_124894.5 NP_200323.1 6 - 7 1462 74
GenBank mRNA AY063788.1 - 6 - 7 1270 74
AY091285.1 - 6 - 7 954 74

View Full Product Details