Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GTE4
Entrez Gene ID
837133
Gene Name
global transcription factor group E4
Gene Aliases
AT1G06230, F9P14.9, F9P14_9, global transcription factor group E4
Chromosome Location
Chr.1: 1906963 - 1911056 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001197993.2 NP_001184922.1 2 - 3 2505 68
NM_001331622.1 NP_001318932.1 2 - 3 2654 68
NM_100505.3 NP_172113.1 2 - 3 2562 68
NM_179270.2 NP_849601.1 1 - 2 2733 68

View Full Product Details