Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CHX6A
Entrez Gene ID
837335
Gene Name
cation/H+ exchanger 6A
Gene Aliases
AT1G08140, ATCHX6A, cation/H+ exchanger 6A
Chromosome Location
Chr.1: 2551935 - 2555229 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_179280.4 NP_849611.1 1 - 2 433 100

View Full Product Details