Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ML5
Entrez Gene ID
839816
Gene Name
MEI2-like protein 5
Gene Aliases
AML5, AT1G29400, F15D2.30, F15D2_30, MEI2-like protein 5
Chromosome Location
Chr.1: 10289998 - 10294630 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001332851.1 NP_001320501.1 10 - 11 2659 71
NM_102680.2 NP_174233.2 10 - 11 2267 71
NM_179396.3 NP_849727.1 10 - 11 2416 71
GenBank mRNA AY062613.1 - 10 - 11 2303 71
AY070368.1 - 10 - 11 2306 71
AY091327.1 - 9 - 10 2143 71

View Full Product Details